Learn More
Leica Biosystems PV POLY-HRP ANTI-RAB IGG 110ML
Catalog No. NC1385426
Encompass
PowerVision Poly-HRP anti-Rabbit IHC Detection Systems are biotin-free, polymeric horseradish peroxidase (HRP) conjugated systems, for the manual detection of tissue-bound rabbit antibodies.
Catalog No. NC1385426 Supplier Leica Biosystems Supplier No. PV6119
PowerVision Poly-HRP anti-Rabbit IHC Detection Systems are biotin-free, polymeric horseradish peroxidase (HRP) conjugated systems, for the manual detection of tissue-bound rabbit antibodies.