Learn More
Cayman Chemical PV9hydrochlorIde 10mg
SDP
Catalog No. 502970929
Encompass_Preferred
A novel α-PVP analog in which the pentyl side chain is elongated by three carbons; intended for forensic and research applications
Catalog No. 50-297-0929 Supplier Cayman Chemical Supplier No. 1506210 MG
A novel α-PVP analog in which the pentyl side chain is elongated by three carbons; intended for forensic and research applications