Learn More
Cayman Chemical PV9 (hydrochloride), Purity: ≥98%, CAS: 2749897-19-4, 50 mg
SDP
Catalog No. 502969606
Encompass_Preferred
A novel α-PVP analog in which the pentyl side chain is elongated by three carbons; intended for forensic and research applications
Catalog No. 50-296-9606 Supplier Cayman Chemical Supplier No. 1506250 MG
A novel α-PVP analog in which the pentyl side chain is elongated by three carbons; intended for forensic and research applications