Learn More
Volu Sol PVA Fixative, Modified (Mercury Free), 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. 50420831 Shop All Volu Sol Products
Encompass_Preferred
Developed as an excellent alternative to mercuric chloride PVA as a fixative for permanent staining.  This item contains no mercury
Catalog No. 50-420-831 Supplier Volu Sol Supplier No. VPM016
Developed as an excellent alternative to mercuric chloride PVA as a fixative for permanent staining.  This item contains no mercury