Learn More
Thermo Scientific™ PVA Modified Fixative Single Vial
Catalog No. R21710 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
12 Vials/Pk.
Catalog No. Quantity
R21710 12 Vials/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R21710 Supplier Thermo Scientific™ Supplier No. R21710
Add to Cart
Add to Cart
This item is not returnable. View return policy
Collect, preserve and permanently stain intestinal parasites with a copper-based fixative, part of system which comes with industry standard color-coding and sporked caps.
Collect, preserve and permanently stain intestinal parasites with copper-based Thermo Scientfic™ PVA Modified Fixative Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Modified PVA Fixatve
Description PVA Modified Fixative Single Vial
Fill Volume 15 mL
Packaging Single vial. Packaged in partitioned boxes.
Quantity 12 Vials/Pk.