Learn More
YMC America, Inc PVA-Sil 120A 5um 50x0.3mm 1/16
SDP
Catalog No. 5000878983
Encompass
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents
Catalog No. 50-008-78983 Supplier YMC America, Inc Supplier No. PV12S0505H0AU
Add to Cart
Add to Cart
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents