Learn More
YMC America, Inc PVA-Sil 120A 5um 75x0.5mm 1/16
SDP
Catalog No. 5000879413
Encompass
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents
Catalog No. 50-008-79413 Supplier YMC America, Inc Supplier No. PV12S05L5J0AU
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents