Learn More
YMC America, Inc PVA-Sil col 120A 5um 100x20mm
SDP
Catalog No. 5000877108
Encompass_Preferred
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents
Catalog No. 50-008-77108 Supplier YMC America, Inc Supplier No. PV12S051020WT
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents