Learn More
YMC America, Inc PVA-Sil guard 120A 5um 10x3mm
SDP
Catalog No. 5000878542
Encompass
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents
Catalog No. 50-008-78542 Supplier YMC America, Inc Supplier No. PV12S050103GC
YMC-Pack PVA-Sil is prepared from a 5 m 120 silica support which is bonded with a monomolecular polymer coating of vinyl alcohol The polymerized PVA completely covers both external and internal surfaces of the silica support protecting it against aggressive high pH buffers and solvents