Learn More
Elemental Scientific Inc PVC 3-stop, Orange/Yellow Stop Colors, Non-Flared, 0.51 mm
SDP
Catalog No. NC1883384
Encompass_Preferred
3-stop tubing is used for pumps that have heads that are smaller than the standard-sized. The colored markers denote the inner diameter size. The markers also hold the tubing in place, stopping it from slipping while the pump is moving.
Catalog No. NC1883384 Supplier Elemental Scientific Inc Supplier No. ES43983051
3-stop tubing is used for pumps that have heads that are smaller than the standard-sized. The colored markers denote the inner diameter size. The markers also hold the tubing in place, stopping it from slipping while the pump is moving.