Learn More
Heathrow Scientific™ PVC Bulk Wrapped Reagent Reservoir
Description
- Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Quality certificate available on request
- Disposable
Specifications
Specifications
| Capacity (English) | 1.7 oz. |
| Color | Clear |
| Capacity (Metric) | 50 mL |
| Dimensions (L x W x H) | 4 x 1.1 x 1.2 in. |
| Material | PVC |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | Reagent Pipetting |
| Packaging | Bulk Wrapped |
| Quantity | 100 Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.