Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ PVC Cleanroom Chemical Aprons with Long Sleeves

Catalog No. 19472670 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-670 Medium
19-472-672 X-Large
19-472-673 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-472-670 Supplier International Enviroguard™ Supplier No. IT6CABMC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Heavy-duty PVC aprons features long sleeves with tunnelized elastic wrists for protection against liquid splash, sprays, and non-hazardous dry particulates.

  • Heavy-duty, 6 mil thick chemical-resistant aprons are made of PVC
  • Designed to protect against acids, alcohol, oils, grease, fats, and a variety of chemicals
  • Liquid-resistant, abrasion-resistant, and puncture-resistant to withstand tough work environments
  • Protect against liquid splash, sprays, and non-hazardous dry particulates
  • Loop over the head and nylon ties on the back
  • Edges are ultrasonically welded to reduce linting and release of unwanted particulates
  • Offer full protection of the front of the body from the chest to the knees or below
  • Long sleeves with tunnelized elastic cuffs to protect the arms from splashes and sprays and to also increase particle containment
  • Cleanroom-ready aprons are clean-processed and individually bagged
  • Applications include semi-conductor, cleanroom, pharmaceutical, chemical processing, petrochemicals, sanitation work
  • Latex-free elastic

Specifications

Type Controlled Environments Apron
Size Medium
Material PVC
Color Blue
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Resistance Liquid, Abrasion, Puncture
Thickness 6 mil
For Use With (Application) Semi-conductor, Cleanroom, Pharmaceutical, Chemical Processing, Petrochemicals, Sanitation Work
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.