Learn More
Research Products International Corp PVC Coated Serum Vial, 15ml, Clear Glass, 60 per Case
SDP
Catalog No. 50550232
Encompass
Specially designed for use with aerosol formulations such as metered dose inhalers for research, development and testing. Vial has a thick glass wall and bottom with a heavy PVC coating for extra protection.
Typical pressure: 2.5 atmosphere
Recommended working capacity: 15 ml
Maximum capacity: 22 ml
Catalog No. 50-550-232 Supplier Research Products International Corp Supplier No. 211065
Specially designed for use with aerosol formulations such as metered dose inhalers for research, development and testing. Vial has a thick glass wall and bottom with a heavy PVC coating for extra protection.
Typical pressure: 2.5 atmosphere
Recommended working capacity: 15 ml
Maximum capacity: 22 ml