Learn More
Research Products International Corp PVC Coated Serum Vial, 360/cs
SDP
Catalog No. 501976909
Encompass
Specially designed for use with aerosol formulations such as metered dose inhalers for research, development and testing. Vial has a thick glass wall and bottom with a heavy PVC coating for extra protection.
Typical pressure: 2.5 atmosphere
Recommended working capacity: 15 ml
Maximum capacity: 22 ml
Catalog No. 50-197-6909 Supplier Research Products International Corp Supplier No. 211068
Specially designed for use with aerosol formulations such as metered dose inhalers for research, development and testing. Vial has a thick glass wall and bottom with a heavy PVC coating for extra protection.
Typical pressure: 2.5 atmosphere
Recommended working capacity: 15 ml
Maximum capacity: 22 ml