Learn More
MSC PVC FLOOR MARKING TAPE
Catalog No. 5000673568
Encompass
This yellow floor marking tape is 3 in. wide by 100 ft. long and constructed from 50 mil thick PVC with a peel-and-stick adhesive backing. It is engineered for heavy-duty applications in high-traffic industrial environments.
  • Does not leave any residue during removal.
  • Warrantied to stay down for 3 years when applied to a clean, dry surface.
  • Allows immediate industrial wheel traffic after application.
  • Beveled edges enable the tape to withstand constant traffic.
  • Easier to install and lasts longer than paint.
  • Easy to clean with any standard floor scrubber, maintaining its color.
Catalog No. 50-006-73568 Supplier MSC Supplier No. 43956804
This yellow floor marking tape is 3 in. wide by 100 ft. long and constructed from 50 mil thick PVC with a peel-and-stick adhesive backing. It is engineered for heavy-duty applications in high-traffic industrial environments.
  • Does not leave any residue during removal.
  • Warrantied to stay down for 3 years when applied to a clean, dry surface.
  • Allows immediate industrial wheel traffic after application.
  • Beveled edges enable the tape to withstand constant traffic.
  • Easier to install and lasts longer than paint.
  • Easy to clean with any standard floor scrubber, maintaining its color.