Learn More
MSC Ergo Advantage Anti-Fatigue Mat, Dry, 18x18x1 in., 1 ea
Catalog No. 5000765533
Encompass
This modular anti-fatigue flooring mat is designed for dry environments. It features a PVC base with raised squares and domes, and includes drainage holes.
  • Interlocking and straight edge design
  • Heavy-duty traffic type
  • PVC base material
  • Surface with raised squares and domes
  • Integrated drainage holes
  • Measures 18 in. long × 18 in. wide × 1 in. thick
Catalog No. 50-007-65533 Supplier MSC Supplier No. 38543880
This modular anti-fatigue flooring mat is designed for dry environments. It features a PVC base with raised squares and domes, and includes drainage holes.
  • Interlocking and straight edge design
  • Heavy-duty traffic type
  • PVC base material
  • Surface with raised squares and domes
  • Integrated drainage holes
  • Measures 18 in. long × 18 in. wide × 1 in. thick