Learn More
MSC Buyers Products Solenoid, 12 Volt, SPST, Continuous Duty, PVC Coated
Supplier: MSC 64148935
This continuous duty solenoid is fully insulated with a PVC coated durable plated steel housing, providing robust protection from the elements. It offers low-input, high-output operation, making it suitable for demanding 12V vehicle applications such as starters, snow plows, lift gates, spreaders, and cranes.
- Fully insulated in PVC coated durable plated steel housing.
- Heavy duty starter keeps your plow or spreader going.
- Low-input, high-output operation.
- 5/16 in.-24 thread terminals for easy installation.
- PVC coated for protection from the elements.
- 85A normally open contacts, SPST, one circuit, off - on.
- Copper studs with 5/16-Inch -24 thread.
- Coil terminals: plated steel, #10-32.
- IP66 rated for protection.
- Flat mount with zinc plated mounting bracket.
- Operating temperature range of -40 degrees C to 85 degrees C.