Learn More
Cole-Parmer Handheld Tubing Cutter, PTFE Coated Steel, Max 1-1/4" OD Tubing
Catalog No. 502557738 Shop All Cole-Parmer Products
Encompass_Preferred
Use tubing cutters to make clean, straight cuts through your plastic tubing. Straight cuts ensure better connection to fittings.
Catalog No. 50-255-7738 Supplier Cole-Parmer Supplier No. 5011850
Use tubing cutters to make clean, straight cuts through your plastic tubing. Straight cuts ensure better connection to fittings.