Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer PVC Lab Tubing, 4 mm ID x 6 mm OD; 15 m

Catalog No. 502557723 Shop All Cole-Parmer Products
Encompass_Preferred

PVC Lab Tubing, 4 mm ID x 6 mm OD; 15 m

Catalog No. 50-255-7723 Supplier Cole-Parmer Supplier No. 5012111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PVC Lab Tubing, 4 mm ID x 6 mm OD; 15 m

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.