Learn More
Thermo Scientific Peristaltic Pump Tubing
Supplier:  Thermo Scientific 1329400
Encompass
Polyvinyl chloride peristaltic pump tubing with 0.254 millimeter internal diameter provides precision flow rates, flexible compression by pump rollers to move fluids, and chemical resistance for analytical applications.
Catalog No. NC1457035