Learn More
Tingley™ PVC on Polyester Rainsuit: Coat only
Catalog No. 19022659 Shop All Tingley Rubber Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-022659 2X-Large
19-022658 Large
19-022656 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-022659 Supplier Tingley™ Supplier No. C53217.2X
This item is not returnable. View return policy
Designed for 100% waterproof protection, these rain coats are generously cut for ease of movement and comfort.
With electronically welded seams, detachable hood, snaps for adjustable fit, and snap front closures.
Specifications
Material PVC, Polyester
Size 2X-Large
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov