Learn More
Thermo Scientific PVC Pump Hose Tubing Pack of 12
Supplier:  Thermo Scientific 1320040
Encompass
Orage/green 0.381 diameter flexible polyvinyl chloride plastic tubing for pumps with good chemical resistance but temperature limitations, not for food contact. Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Catalog No. NC0841501