Learn More
Glass Expansion PVC Pump Tube 2tag 1.02mm ID White/White, PKT 12, (3710034400, 5000-090, 5064-8028)
SDP
Supplier:  Glass Expansion 1.02WHT
Encompass_Preferred
PVC Pump Tube 2tag 1.02mm ID White/White (PKT 12).Tag Colors: White/WhiteID mm: 1.02Wall Thickness mm: 0.86OD mm: 2.74Overall Length: 455mm (152mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Thermoplastic, PVC based material with plasticizerSpecial Properties: Inexpensive all-around tubing for general laboratory applicationsChemical Compatibility: Excellent for aqueous solutions of the following acids and concentrations, up to 35% Nitric Acid, up to 37% Hydrochloric Acid, up to 25% Hydrofluoric Acid, up to 30% Hydrogen Peroxide. For >35% Nitric Acid and >25% Hydrofluoric Acid we recommend using Viton P/N VS1.02-WHT.Limitations: Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1325793