Learn More
Perkin Elmer US LLC PVC Pure Flared Two-Stop Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. 502103842
Encompass_Preferred
This flared end PVC Pure Pump Tubing is for use with NexION 1000 and 2000 ICP-MS series instruments. It is the standard two-stop tubing for NexION 1000/2000 and ships as a package of 12.
Catalog No. 50-210-3842 Supplier Perkin Elmer US LLC Supplier No. N8152407
This flared end PVC Pure Pump Tubing is for use with NexION 1000 and 2000 ICP-MS series instruments. It is the standard two-stop tubing for NexION 1000/2000 and ships as a package of 12.