Learn More
Perkin Elmer US LLC PVC Pure Flared Two-Stop Pump Tubing - 0.51 mm I.D., Orange/Yellow, Pkg. 12
Catalog No. 502103841
Encompass_Preferred
This flared end PVC Pure Pump Tubing is for use with NexION 1000 and 2000 ICP-MS series instruments. It is the standard two-stop tubing for NexION 1000/2000 and ships as a package of 12.
Catalog No. 50-210-3841 Supplier Perkin Elmer US LLC Supplier No. N8152405
This flared end PVC Pure Pump Tubing is for use with NexION 1000 and 2000 ICP-MS series instruments. It is the standard two-stop tubing for NexION 1000/2000 and ships as a package of 12.