Learn More
Electron Microscopy Sciences PVC Sample Bottles 50 ml, 36 mm Neck opening, 60 mm height
SDP
Catalog No. 5028418
Encompass_Preferred
These lightweight transparent PVC sample bottles come with snap-on tight-fitting caps and they offer excellent clarity, durability, and good chemical resistance. Maximum temperature -20 degree C (-4 degree F) to 80 degree C (176 degree F).
Neck opening: 36 mm
Catalog No. 50-284-18 Supplier Electron Microscopy Sciences Supplier No. 6426405
These lightweight transparent PVC sample bottles come with snap-on tight-fitting caps and they offer excellent clarity, durability, and good chemical resistance. Maximum temperature -20 degree C (-4 degree F) to 80 degree C (176 degree F).
Neck opening: 36 mm