Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, PVC Solvent/Hydrocarbon, 1.30 mm ID; 12/PK
Catalog No. NC0131342
Encompass
Appearance: Yellow, translucent Temperature: ¿30 to 85degC Life at 100 rpm:1000 hrs Advantages: Resistant to ozone and UV light; resistant to cracking, swelling, and hardening. Applications: Ideal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolants.
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC0131342 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9560532
Appearance: Yellow, translucent Temperature: ¿30 to 85degC Life at 100 rpm:1000 hrs Advantages: Resistant to ozone and UV light; resistant to cracking, swelling, and hardening. Applications: Ideal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolants.
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.