Learn More
MSC PRO-SAFE Anti-Fatigue Mat, Urethane, 3/8 in. Thick, 1 ea.
Catalog No. 5000766020
Encompass
This urethane-coated vinyl sponge anti-fatigue mat is designed for use in dry work areas. The urethane coating provides the comfort of sponge matting with a chemical and abrasion-resistant surface, making it ideal for medium traffic areas.
  • Features beveled edges and a pebbled surface
  • Suitable for medium-duty traffic in dry work areas
  • Urethane coating provides comfort of sponge matting with chemical and abrasion resistance
Catalog No. 50-007-66020 Supplier MSC Supplier No. 88266507
This urethane-coated vinyl sponge anti-fatigue mat is designed for use in dry work areas. The urethane coating provides the comfort of sponge matting with a chemical and abrasion-resistant surface, making it ideal for medium traffic areas.
  • Features beveled edges and a pebbled surface
  • Suitable for medium-duty traffic in dry work areas
  • Urethane coating provides comfort of sponge matting with chemical and abrasion resistance