Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PVC Reinforced, 1/2"" ID x 3/4"" OD; 100 ft
Catalog No. NC1629209
Encompass
Applications: High-pressure use, food processing, and beverage handling. Characteristics: Good acid/alkali resistance and aging characteristics. White polyester yarn with orange tracer yarn embedded in a transparent PVC wall. Certification: Resin is FDA-compliant (21 CFR 177.199) and meets USDA requirements. NSF-listed (standard 51) with 3A Sanitary Standards for dairy. Temperature range: -5 to 180¿F (-21 to 82¿C). NSF temperature to 140¿F. Sterilize: Ethylene oxide.
Catalog No. NC1629209 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0660216
Applications: High-pressure use, food processing, and beverage handling. Characteristics: Good acid/alkali resistance and aging characteristics. White polyester yarn with orange tracer yarn embedded in a transparent PVC wall. Certification: Resin is FDA-compliant (21 CFR 177.199) and meets USDA requirements. NSF-listed (standard 51) with 3A Sanitary Standards for dairy. Temperature range: -5 to 180¿F (-21 to 82¿C). NSF temperature to 140¿F. Sterilize: Ethylene oxide.