Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, PVC, 1.52 mm ID; 12/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9562536
Encompass
Three-Stop PVC Color-coded Autoanalysis Tubing for Ismatec Pumps is USP Class VI tubing that is Phthalate-free. It has a long pumping life and exhibits a wide range of acid and chemical resistance, thus making it an ideal choice for Pharmaceutical industries. Plasticizer is very tightly bonded into the material, preventing any leaching of component chemicals into the medium being transported, maintaining its sterility. The tubing is transparent, which allows visual observation of tubing contents. It also offers very low gas permeability and can withstand high pressure contents.
Tubing is also called TAAT / microbore and auto analysis.
The overall length of Lab PVC 95625-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1145879