Learn More
AVANTOR MASTERFLEX (VWR LLC) Plastic luer fittings, PVC tubing with luer ends, 24"" L, M/M fitting ends
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3060062
Encompass
Plastic luer fittings, PVC tubing with luer ends, 24" L, M/M fitting ends, High pressure tubing has luer fittings at each end¿ male luer lok x female luer in varying lengths from 10" to 48". Can be sterilized by ETO.Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Catalog No. NC1376778