Learn More
ATCC pVC-604 plasmid; in E. coli
Supplier:  ATCC MBA212
Encompass
pVC-604 plasmid in E. coli is a vector that can be used with transformation-associated recombination (TAR) in yeast.
Catalog No. 50-238-1968