Learn More
CELLTREAT Scientific Products PVDF Syringe Filter, 0.10m, 30mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229741
Encompass_Preferred
CELLTREAT Scientific Products 0.10¿m PVDF Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. PVDF (Polyvinylidene Flouride). Low protein-binding for filtration of non-aggressive aqueous and mild organic solutions. Maximizes protein recovery. High temperature filtration. Membrane diameter 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually packed in peel-to-open paper/plastic blister packs, 30/case.
Catalog No. 50-202-056