Learn More
CELLTREAT Scientific Products PVDF Syringe Filter, 0.22m, 13mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229742
Encompass_Preferred
CELLTREAT Scientific Products 0.22¿m PVDF Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. PVDF (Polyvinylidene Flouride). Low protein-binding for filtration of non-aggressive aqueous and mild organic solutions. Maximizes protein recovery. High temperature filtration. Membrane size 13mm. RNase/DNase free and non-pyrogenic.Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister packs. 75/case.
Catalog No. 50-202-057