Learn More
Cole-Parmer Essentials PVDF Chromatography Syringe Filters; 0.2µm Pore, 13mm Dia, 100/Pk
Supplier:  Cole-Parmer 1594516
Encompass_Preferred
PVDF Syringe Filters are both low protein binding and possess high flow rates, and offer great chemical compatibility—making them ideal for solutions that are difficult to filter. Best suited for applications requiring sterilization and clarification of biological solutions, and preparing HPLC solutions and aqueous samples. Maximum use temperature is 60°C (140°F).
Catalog No. NC2177905