Learn More
Bio Rad Laboratories PVDF MEMBRANE SHEETS 7X8.4CM
Catalog No. NC9477962
Encompass
Immun-Blot PVDF Membrane, 10 sheets, 7 x 8.4cm, For immunoblotting of transferred proteins, high capacity and low background, Holds up under repeated stripping and reprobing applications
Catalog No. NC9477962 Supplier Bio Rad Laboratories Supplier No. 1620174
Immun-Blot PVDF Membrane, 10 sheets, 7 x 8.4cm, For immunoblotting of transferred proteins, high capacity and low background, Holds up under repeated stripping and reprobing applications