Learn More
Electron Microscopy Sciences PVDF Plastic Probes Kit
SDP
Catalog No. 501930657
Encompass_Preferred
Magic Wands are made from PEEK, a high performance plastic, with excellent chemical resistantance and withstands high temperatures (up to 300 degree C). Magic wand is ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or evendosing samples etc.
Catalog No. 50-193-0657 Supplier Electron Microscopy Sciences Supplier No. 6332304
Magic Wands are made from PEEK, a high performance plastic, with excellent chemical resistantance and withstands high temperatures (up to 300 degree C). Magic wand is ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or evendosing samples etc.