Learn More
Enzo Life Sciences PVDF Transfer Membrane (7cm x 9cm) (10 ea)
SDP
Supplier:  Enzo Life Sciences ENZKIT1640001
Encompass_Preferred
PVDF membranes provide high surface area for strong hydrophobic interactions. PVDF membranes typically absorb 50% more protein than nylon or nitrocellulose membranes. This membrane has been optimized for Western transfer applications compatible with WESTERNVIEW™ Detection Kits or other chemiluminescent or chromogenic Western blot detection substrates. Long Term Storage: . Applications: WB .
Catalog No. 50-201-3854