Learn More
Accuflow Systems Saint-Gobain BarbLock® Tubing Retainers
Supplier:  Accuflow Systems BL135254ALLRRPVDF
Encompass_Preferred
Saint-Gobain BarbLock® tubing retainers are designed to secure flexible tubing and hose onto barbed fittings. They feature a full 360° radial crimp behind the barb and 360° of compression directly over the barb's ridge to prevent leaks.
  • Constructed from high-performance PVDF resin
  • Meets USP <88> Class VI, USP <87>, and/or ISO 10993-5 requirements
  • Gamma stable
Catalog No. NC2868127