Learn More
Sigma Aldrich Fine Chemicals Biosciences PVDf Western Blotting Memb
Catalog No. 502906116
Encompass
PVDf Western Blotting Membranes are hydrophobic microporous polyvinylidene difluoride (PVDf) membranes with a pore size of 0.2mum. The PVDf Western Blotting Membranes are ideal for western and dot blots. The high binding capacity high mechanical strength and chemical resistance of the PVDf membrane makes it specifically useful for protein blotting. The combination of strong sample retention and low background binding generates excellent signal-to-noise ratios in colorimetric and chemiluminescent detection. The primary binding mechanism includes electrostatic and hydrophobic nature. Adsorption Goat IgG 294mug/cm2 BSA 131mug/cm2 insulin 85mug/cm2
Catalog No. 50-290-6116 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3010040001
PVDf Western Blotting Membranes are hydrophobic microporous polyvinylidene difluoride (PVDf) membranes with a pore size of 0.2mum. The PVDf Western Blotting Membranes are ideal for western and dot blots. The high binding capacity high mechanical strength and chemical resistance of the PVDf membrane makes it specifically useful for protein blotting. The combination of strong sample retention and low background binding generates excellent signal-to-noise ratios in colorimetric and chemiluminescent detection. The primary binding mechanism includes electrostatic and hydrophobic nature. Adsorption Goat IgG 294mug/cm2 BSA 131mug/cm2 insulin 85mug/cm2