Learn More
Med Vet International Povidone Iodine Prep Pads, 1000/Case
Catalog No. 501180483
Encompass_Preferred
Povidone Iodine Prep Pads: Povidone Iodine is a broad-range antimicrobial that kills bacteria, fungi, viruses, protozoa and yeast. Forms skin protection against microbes until the solution is washed from the skin. Each pad or swabstick is saturated with a 10% PVP-I, USP solution equivalent to 1% available iodine. Prep Pad, Medium. Compared to Betadine.
Catalog No. 50-118-0483 Supplier Med Vet International Supplier No. MDS093917
Povidone Iodine Prep Pads: Povidone Iodine is a broad-range antimicrobial that kills bacteria, fungi, viruses, protozoa and yeast. Forms skin protection against microbes until the solution is washed from the skin. Each pad or swabstick is saturated with a 10% PVP-I, USP solution equivalent to 1% available iodine. Prep Pad, Medium. Compared to Betadine.