Learn More
Aviva Systems Biology Corp PVR Antibody - N-terminal region (ARP63030_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PVR. It was validated on Western Blot. . Alternate names: CD155, FLJ25946, HVED, NECL5, Necl-5, PVS, TAGE4ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-287-849