Learn More
Medchemexpress LLC HY-P7401 10ug Medchemexpress, PVR/CD155, Fc Chimera (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP740110UG
Encompass_Preferred
Medchemexpress, HY-P7401 10ug PVR/CD155, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human PVR/CD155, Fc Chimera (HEK293-expressed), a Type I transmembrane glycoprotein in the immunoglobulin superfamily, is involved in the cellular poliovirus infection in primates and intestinal humoral immune responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3159