Learn More
Aviva Systems Biology Corp PVRIG Antibody - middle region (ARP49798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PVRIG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PVRIG. Alternate names: C7orf15, MGC104322, MGC138295, MGC138297, MGC2463ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60187