Learn More
ACROBiosystems ACROBiosystems Mouse PVRIG Protein, Fc Tag
Supplier:  ACROBiosystems PVGM52571MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse PVRIG, Fc Tag (PVG-M5257) is expressed from human 293 cells (HEK293). It contains AA Ser 35 - Asp 165 (Accession # A0A1B0GS01-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6914