Learn More
ACROBiosystems ACROBiosystems Human PVRIG Protein, Fc Tag
Catalog No. 501956849
Encompass_Preferred
Recombinant Protein;1MG;Human PVRIG, Fc Tag (PVG-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.1 kDa. As a result of glycosylation, the protein migrates as 50 kDa under reducing (R) condition, and 90 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 μg/mL (100 μL/well) can bind Human PVRIG, Fc Tag (Cat. No. PVG-H5257) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-6849 Supplier ACROBiosystems Supplier No. PVGH52571MG
Recombinant Protein;1MG;Human PVRIG, Fc Tag (PVG-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.1 kDa. As a result of glycosylation, the protein migrates as 50 kDa under reducing (R) condition, and 90 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 5 μg/mL (100 μL/well) can bind Human PVRIG, Fc Tag (Cat. No. PVG-H5257) with a linear range of 2-31 ng/mL (QC tested).