Learn More
ACROBiosystems ACROBiosystems Human PVRIG Protein, His Tag
Supplier:  ACROBiosystems PVGH52H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human PVRIG, His Tag (PVG-H52H4) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa. The protein migrates as 20-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PVRIG, His Tag (Cat. No. PVG-H52H4) at 2 μg/mL (100 μL/well) can bind Human Nectin-2, Fc Tag (Cat. No. PV2-H5253) with a linear range of 0.5-8 ng/mL (QC tested).;Immobilized Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) at 1 μg/mL (100 μL/well) can bind Human PVRIG, His Tag (Cat. No. PVG-H52H4) with a linear range of 1-39 ng/mL (Routinely tested).
Catalog No. 50-195-7204