Learn More
Aviva Systems Biology Corp NECTIN3 Antibody - middle region (ARP42447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PVRL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PVRL3. Alternate names: CDw113, DKFZP566B0846, FLJ90624, PPR3, PRR3, PVRR3, nectin-3, CD113, CDW113, NECTIN-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-002