Learn More
Aviva Systems Biology Corp NECTIN3 Antibody - N-terminal region (ARP42446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PVRL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PVRL3. Alternate names: CDw113, DKFZP566B0846, FLJ90624, PPR3, PRR3, PVRR3, nectin-3, CD113, CDW113, NECTIN-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58227