Learn More
OriGene PVRL4 (NECTIN4) (NM_030916) Human Over-expression Lysate, 100ug
SDP
Catalog No. NC3198534
Encompass_Preferred
Components:1 vial of 100 µg gene specific transient over-expression cell lysate in RIPA buffer1 vial of 100 µg whole HEK293T cell lysate in RIPA buffer1 vial of 250ul 2xSDS Sample Buffer (4% SDS, 125mM Tris-HCl pH6.8, 10% Glycerol, 0.002% Bromophenol blue, 100mM DTT)
Catalog No. NC3198534 Supplier OriGene Supplier No. LY403093
Components:1 vial of 100 µg gene specific transient over-expression cell lysate in RIPA buffer1 vial of 100 µg whole HEK293T cell lysate in RIPA buffer1 vial of 250ul 2xSDS Sample Buffer (4% SDS, 125mM Tris-HCl pH6.8, 10% Glycerol, 0.002% Bromophenol blue, 100mM DTT)